3-Cyanpyridin-N-oxid 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD3284-5GEA 126 EUR
BLDPBD3284-5G BLDPBD3284-25G BLDPBD3284-1G
3-Cyanpyridin-N-oxid 98%
3-Cyanpyridin-N-oxid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₄N₂O
Molecular Weight: 120,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00129037
CAS-Nummer: 14906-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR