3-Chlor-5-fluorphenol 98%

BLDPBD32554-10GEA 121 EUR
BLDPBD32554-10G BLDPBD32554-25G BLDPBD32554-1G BLDPBD32554-100G BLDPBD32554-5G
3-Chlor-5-fluorphenol 98%
3-Chlor-5-fluorphenol

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₄ClFO
Molecular Weight: 146,55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00042212
CAS-Nummer: 202982-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR