4-(Piperidin-4-yl)benzonitrile hydrochloride 95%

BLDPBD32176-1GEA 579 EUR
BLDPBD32176-1G BLDPBD32176-50MG BLDPBD32176-100MG BLDPBD32176-250MG
4-(Piperidin-4-yl)benzonitrile hydrochloride 95%
4-(Piperidin-4-yl)benzonitrile hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₂H₁₄N₂·ClH
Molecular Weight: 222,714 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08544124
CAS-Nummer: 162997-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR