4-Fluor-2-(trifluormethoxy)benzylbromid 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD321133-1GEA 158 EUR
BLDPBD321133-1G BLDPBD321133-5G
4-Fluor-2-(trifluormethoxy)benzylbromid 95%
4-Fluor-2-(trifluormethoxy)benzylbromid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₈H₅BrF₄O
Molecular Weight: 273,02 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD19687175
CAS-Nummer: 1323966-22-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR