2-Fluor-4-methoxybenzylamin 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD318117-250MGEA 240 EUR
BLDPBD318117-250MG BLDPBD318117-1G BLDPBD318117-100MG
2-Fluor-4-methoxybenzylamin 95%
2-Fluor-4-methoxybenzylamin

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₈H₁₀FNO
Molecular Weight: 155,1700 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04116358
CAS-Nummer: 937843-59-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR