1-(2-Methoxyethyl)piperazin 96%

BLDPBD31798-5GEA 27 EUR
BLDPBD31798-5G BLDPBD31798-25G BLDPBD31798-100G BLDPBD31798-250MG BLDPBD31798-10G BLDPBD31798-1G
1-(2-Methoxyethyl)piperazin 96%
1-(2-Methoxyethyl)piperazin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₁₆N₂O
Molecular Weight: 144,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00191214
CAS-Nummer: 13484-40-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR