6-Bromo-1,5-dimethyl-1H-indazole 96%

Lieferant: BLD PHARMATECH GMBH

BLDPBD317003-100MGEA 49 EUR
BLDPBD317003-100MG BLDPBD317003-250MG BLDPBD317003-1G
6-Bromo-1,5-dimethyl-1H-indazole 96%
6-Bromo-1,5-dimethyl-1H-indazole

Storage condition: Sealed in dry, 2-8°C

Formel: C₉H₉BrN₂
Molecular Weight: 225,085 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12028628
CAS-Nummer: 1159511-83-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR