(3,5-Difluorphenyl)methansulfonsäurefluorid 95%

BLDPBD315365-1GEA 86 EUR
BLDPBD315365-1G BLDPBD315365-5G BLDPBD315365-250MG
(3,5-Difluorphenyl)methansulfonsäurefluorid 95%
(3,5-Difluorphenyl)methansulfonsäurefluorid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₇H₅ClF₂O₂S
Molecular Weight: 226,63 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD09743349
CAS-Nummer: 163295-74-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR