Kalium(3-iodphenyl)trifluoroborat 98+%

Lieferant: BLD PHARMATECH GMBH

BLDPBD314325-5GEA 205 EUR
BLDPBD314325-5G BLDPBD314325-250MG BLDPBD314325-1G
Kalium(3-iodphenyl)trifluoroborat 98+%
Potassium-3-iodophenyltrifluoroborate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₄BIF₃K
Molecular Weight: 309,91 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800739
CAS-Nummer: 1189097-36-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR