Tasquinimod 98%

BLDPBD305716-5MGEA 125 EUR
BLDPBD305716-5MG
Tasquinimod 98%
Tasquinimod

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₂₀H₁₇F₃N₂O₄
Molecular Weight: 406,36 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18251454
CAS-Nummer: 254964-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR