2-(2-Bromophenyl)-4,6-diphenyl-1,3,5-triazine 99%
|
|
BLDPBD304988-5GEA
65
EUR
BLDPBD304988-5G
BLDPBD304988-10G
BLDPBD304988-25G
BLDPBD304988-1G
2-(2-Bromophenyl)-4,6-diphenyl-1,3,5-triazine 99%
2-(2-Bromophenyl)-4,6-diphenyl-1,3,5-triazine
Storage condition: Sealed in dry, Room Temperature
Formel:
C₂₁H₁₄BrN₃ Molecular Weight: 388,27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29047055 CAS-Nummer: 77989-15-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...