3,4-(Methylenedioxy)phenylboronsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD29929-250MGEA 6 EUR
BLDPBD29929-250MG BLDPBD29929-1G BLDPBD29929-100G BLDPBD29929-25G BLDPBD29929-5G BLDPBD29929-10G
3,4-(Methylenedioxy)phenylboronsäure 98%
3,4-(Methylenedioxy)phenylboronsäure

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₇BO₄
Molecular Weight: 165,94 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01009695
CAS-Nummer: 94839-07-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR