1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulphonyl)ethanamine 97%

BLDPBD297608-25GEA 79 EUR
BLDPBD297608-25G BLDPBD297608-1G BLDPBD297608-250MG BLDPBD297608-5G BLDPBD297608-10G
1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulphonyl)ethanamine 97%
1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulphonyl)ethanamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₂H₁₉NO₄S
Molecular Weight: 273,35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27665207
CAS-Nummer: 253168-94-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR