Bis[(pinacolato)boryl]methane 95%

BLDPBD293619-5GEA 82 EUR
BLDPBD293619-5G BLDPBD293619-0.025 BLDPBD293619-10G BLDPBD293619-100G BLDPBD293619-25G BLDPBD293619-1G
Bis[(pinacolato)boryl]methane 95%
Bis[(pinacolato)boryl]methane

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₃H₂₆B₂O₄
Molecular Weight: 267,97 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27977747
CAS-Nummer: 78782-17-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR