5,6-Difluoro-2,1,3-benzothiadiazole 95%

BLDPBD292641-100MGEA 13 EUR
BLDPBD292641-100MG BLDPBD292641-250MG BLDPBD292641-1G BLDPBD292641-5G
5,6-Difluoro-2,1,3-benzothiadiazole 95%
5,6-Difluoro-2,1,3-benzothiadiazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₆H₂F₂N₂S
Molecular Weight: 172,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28100486
CAS-Nummer: 1293389-28-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR