2-[(6-Chloropyridin-3-yl)methyl]-2,3-dihydro-1H-isoindole-1,3-dione 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD288452-250MGEA 45 EUR
BLDPBD288452-250MG BLDPBD288452-1G BLDPBD288452-5G BLDPBD288452-100MG
2-[(6-Chloropyridin-3-yl)methyl]-2,3-dihydro-1H-isoindole-1,3-dione 97%
2-[(6-Chloropyridin-3-yl)methyl]-2,3-dihydro-1H-isoindole-1,3-dione

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₄H₉ClN₂O₂
Molecular Weight: 272,686 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09802551
CAS-Nummer: 120739-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR