Benzyldimethyltetradecylammoniumchlorid Dihydrat 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD287644-5GEA 7 EUR
BLDPBD287644-5G BLDPBD287644-25G BLDPBD287644-1G
Benzyldimethyltetradecylammoniumchlorid Dihydrat 98%
Benzyldimethyltetradecylammoniumchlorid Dihydrat

Storage condition: Inert atmosphere, Room Temperature

Formel: C₂₃H₄₂ClN·2H₂O
Molecular Weight: 404,0699 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00149985
CAS-Nummer: 147228-81-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR