(1H-Pyrrolo[2,3-c]pyridin-2-yl)methanol 95%

BLDPBD285572-1GEA 471 EUR
BLDPBD285572-1G BLDPBD285572-250MG BLDPBD285572-100MG
(1H-Pyrrolo[2,3-c]pyridin-2-yl)methanol 95%
(1H-Pyrrolo[2,3-c]pyridin-2-yl)methanol

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₈N₂O
Molecular Weight: 148,162 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11506123
CAS-Nummer: 882881-15-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR