2-Methyl-4-thiazolcarbonsäure 97%
|
|
BLDPBD28472-5GEA
33
EUR
BLDPBD28472-5G
BLDPBD28472-1G
BLDPBD28472-10G
BLDPBD28472-100G
BLDPBD28472-250MG
BLDPBD28472-25G
2-Methyl-4-thiazolcarbonsäure 97%
2-Methyl-4-thiazolecarboxylic acid
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₅H₅NO₂S Molecular Weight: 143,1637 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03407332 CAS-Nummer: 35272-15-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...