5-(2-Pyridyl)-1H-tetrazol 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD28418-250MGEA 9 EUR
BLDPBD28418-250MG BLDPBD28418-25G BLDPBD28418-5G BLDPBD28418-1G
5-(2-Pyridyl)-1H-tetrazol 98%
5-(2-Pyridyl)-1H-tetrazole

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₅N₅
Molecular Weight: 147,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00068114
CAS-Nummer: 33893-89-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR