3-Amino-4-methoxypyridin 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD28410-25GEA 416 EUR
BLDPBD28410-25G BLDPBD28410-100MG BLDPBD28410-1G BLDPBD28410-10G BLDPBD28410-250MG BLDPBD28410-5G
3-Amino-4-methoxypyridin 98%
3-Amino-4-methoxypyridine

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formel: C₆H₈N₂O
Molecular Weight: 124,1400 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD07374959
CAS-Nummer: 33631-09-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR