3-Bromo-9-(2-naphthyl)carbazole 97%

BLDPBD280817-100GEA 555 EUR
BLDPBD280817-100G BLDPBD280817-25G BLDPBD280817-5G BLDPBD280817-1G
3-Bromo-9-(2-naphthyl)carbazole 97%
3-Bromo-9-(2-naphthyl)carbazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₂H₁₄BrN
Molecular Weight: 372,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26127422
CAS-Nummer: 934545-80-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR