4-Chlor-2-fluor-3-methoxybenzoesäure 95%

BLDPBD280110-5GEA 270 EUR
BLDPBD280110-5G BLDPBD280110-1G BLDPBD280110-250MG
4-Chlor-2-fluor-3-methoxybenzoesäure 95%
4-Chloro-2-fluoro-3-methoxybenzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₆ClFO₃
Molecular Weight: 204,59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19687204
CAS-Nummer: 1169870-80-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR