1-Boc-piperidin-4-ylidenessigsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD27916-10GEA 119 EUR
BLDPBD27916-10G BLDPBD27916-5G BLDPBD27916-25G BLDPBD27916-1G
1-Boc-piperidin-4-ylidenessigsäure 98%
1-Boc-piperidin-4-ylidenessigsäure

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₂H₁₉NO₄
Molecular Weight: 241,2836 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04115019
CAS-Nummer: 193085-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR