5-Bromo-1-methylisatin 95%

BLDPBD278248-25GEA 588 EUR
BLDPBD278248-25G BLDPBD278248-5G BLDPBD278248-250MG BLDPBD278248-100MG BLDPBD278248-1G
5-Bromo-1-methylisatin 95%
5-Bromo-1-methylisatin

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₆BrNO₂
Molecular Weight: 240,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00456313
CAS-Nummer: 2058-72-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR