4-Brom-3,5-dimethyl-1H-pyrazol 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD2778-25GEA 19 EUR
BLDPBD2778-25G BLDPBD2778-500G BLDPBD2778-100G BLDPBD2778-5G
4-Brom-3,5-dimethyl-1H-pyrazol 95%
4-Bromo-3,5-dimethyl-1H-pyrazole

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₅H₇BrN₂
Molecular Weight: 175,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005242
CAS-Nummer: 3398-16-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR