Methyl-1H-indazol-6-carboxylat 97%

BLDPBD27740-25GEA 74 EUR
BLDPBD27740-25G BLDPBD27740-250MG BLDPBD27740-1G BLDPBD27740-500G BLDPBD27740-10G BLDPBD27740-100G BLDPBD27740-5G
Methyl-1H-indazol-6-carboxylat 97%
Methyl-1H-indazol-6-carboxylat

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₈N₂O₂
Molecular Weight: 176,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07371612
CAS-Nummer: 170487-40-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR