4-Bromo-3-phenylpyrazole 98%

BLDPBD27487-100MGEA 7 EUR
BLDPBD27487-0.025G BLDPBD27487-100MG BLDPBD27487-1G BLDPBD27487-250MG BLDPBD27487-5G BLDPBD27487-25G
4-Bromo-3-phenylpyrazole 98%
4-Bromo-3-phenylpyrazole

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₇BrN₂
Molecular Weight: 223,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09027561
CAS-Nummer: 13808-65-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR