N-[(3-Pyridyl)methyl]-2-propanamine 95%

BLDPBD274717-5GEA 484 EUR
BLDPBD274717-5G BLDPBD274717-1G BLDPBD274717-250MG
N-[(3-Pyridyl)methyl]-2-propanamine 95%
N-[(3-Pyridyl)methyl]-2-propanamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₁₄N₂
Molecular Weight: 150,221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00716868
CAS-Nummer: 19730-12-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR