4,5-Dimethoxy-2-methylbenzoic acid 97%

BLDPBD273502-250MGEA 69 EUR
BLDPBD273502-250MG BLDPBD273502-5G BLDPBD273502-100MG BLDPBD273502-1G
4,5-Dimethoxy-2-methylbenzoic acid 97%
4,5-Dimethoxy-2-methylbenzoic acid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₀H₁₂O₄
Molecular Weight: 196,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00029889
CAS-Nummer: 20736-28-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR