2-Brom-3-nitrophenol 98%

BLDPBD27124-100MGEA 10 EUR
BLDPBD27124-100MG BLDPBD27124-250MG BLDPBD27124-10G BLDPBD27124-5G BLDPBD27124-1G
2-Brom-3-nitrophenol 98%
2-Brom-3-nitrophenol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₄BrNO₃
Molecular Weight: 218,0100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08704555
CAS-Nummer: 101935-40-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR