2-Hydroxy-5-(pyridin-3-yl)benzaldehyde 95%

BLDPBD271073-1GEA 438 EUR
BLDPBD271073-1G BLDPBD271073-250MG BLDPBD271073-100MG
2-Hydroxy-5-(pyridin-3-yl)benzaldehyde 95%
2-Hydroxy-5-(pyridin-3-yl)benzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₂H₉NO₂
Molecular Weight: 199,205 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11877750
CAS-Nummer: 85633-41-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR