4-Iod-1-methylimidazol-5-carbaldehyd 95+%

Lieferant: BLD PHARMATECH GMBH

BLDPBD2683-250MGEA 134 EUR
BLDPBD2683-250MG BLDPBD2683-100MG
4-Iod-1-methylimidazol-5-carbaldehyd 95+%
4-Iod-1-methylimidazol-5-carbaldehyd

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₅H₅IN₂O
Molecular Weight: 236,0105 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01632219
CAS-Nummer: 189014-13-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR