Mal-amido-PEG2-NHS 95%
|
|
BLDPBD268025-1GEA
366
EUR
BLDPBD268025-1G
BLDPBD268025-250MG
BLDPBD268025-100MG
Mal-amido-PEG2-NHS 95%
Mal-amido-PEG2-NHS
Storage condition: Inert atmosphere, Store in freezer, under -20°C
Formel:
C₁₈H₂₃N₃O₉ Molecular Weight: 425,39 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD11041137 CAS-Nummer: 955094-26-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...