5-Fluor-3-pyridinboronsäure 95%
|
|
BLDPBD2641-10GEA
134
EUR
BLDPBD2641-10G
BLDPBD2641-1G
BLDPBD2641-25G
BLDPBD2641-5G
BLDPBD2641-250MG
5-Fluor-3-pyridinboronsäure 95%
5-Fluoro-3-pyridineboronic acid
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₅H₅BFNO₂ Molecular Weight: 140,91 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368243 CAS-Nummer: 872041-86-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...