4'-Brom-3'-methylacetophenon 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD263114-1GEA 198 EUR
BLDPBD263114-1G BLDPBD263114-5G BLDPBD263114-250MG BLDPBD263114-100MG
4'-Brom-3'-methylacetophenon 95%
4'-Brom-3'-methylacetophenon

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₉BrO
Molecular Weight: 229,0706 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17169058
CAS-Nummer: 50870-44-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR