5-Brom-2,4-difluorbenzaldehyd 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD262950-100GEA 561 EUR
BLDPBD262950-100G BLDPBD262950-25G BLDPBD262950-1G BLDPBD262950-5G BLDPBD262950-250MG BLDPBD262950-10G
5-Brom-2,4-difluorbenzaldehyd 98%
5-Brom-2,4-difluorbenzaldehyd

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₃BrF₂O
Molecular Weight: 220,9989 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09842439
CAS-Nummer: 473416-91-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR