2-Amino-4,5-dimethylbenzoic acid 97%

BLDPBD262944-1GEA 29 EUR
BLDPBD262944-1G BLDPBD262944-250MG BLDPBD262944-100MG
2-Amino-4,5-dimethylbenzoic acid 97%
2-Amino-4,5-dimethylbenzoic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00156998
CAS-Nummer: 15089-51-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR