2-(2-Biphenylyl)amino-9,9-dimethylfluorene 97%

BLDPBD262479-100MGEA 14 EUR
BLDPBD262479-100MG BLDPBD262479-1G BLDPBD262479-250MG BLDPBD262479-25G BLDPBD262479-5G
2-(2-Biphenylyl)amino-9,9-dimethylfluorene 97%
2-(2-Biphenylyl)amino-9,9-dimethylfluorene

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₂₇H₂₃N
Molecular Weight: 361,49 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19440968
CAS-Nummer: 1198395-24-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR