Boc-(R)-3-Cyclopentylalanine 95%
|
|
BLDPBD261493-1GEA
184
EUR
BLDPBD261493-1G
BLDPBD261493-100MG
BLDPBD261493-250MG
Boc-(R)-3-Cyclopentylalanine 95%
Boc-(R)-3-Cyclopentylalanine
Storage condition: Keep in dark place, Sealed in dry, 2-8°C
Formel:
C₁₃H₂₃NO₄ Molecular Weight: 257,326 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD03094882 CAS-Nummer: 219819-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...