2-Ethoxy-3-formyl-5-methylphenylboronsäure 95%
|
|
BLDPBD261012-5GEA
240
EUR
BLDPBD261012-5G
BLDPBD261012-1G
BLDPBD261012-250MG
2-Ethoxy-3-formyl-5-methylphenylboronsäure 95%
2-Ethoxy-3-formyl-5-methylphenylboronsäure
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₁₀H₁₃BO₄ Molecular Weight: 208,0200 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD08276749 CAS-Nummer: 480424-54-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...