3-Bromo-8-methylimidazo[1,2-a]pyridine 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD259427-1GEA 216 EUR
BLDPBD259427-1G BLDPBD259427-250MG BLDPBD259427-100MG
3-Bromo-8-methylimidazo[1,2-a]pyridine 95%
3-Bromo-8-methylimidazo[1,2-a]pyridine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₇BrN₂
Molecular Weight: 211,059 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06496194
CAS-Nummer: 866135-66-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR