2-Iodo-6-nitrophenol 97%

BLDPBD259064-100MGEA 68 EUR
BLDPBD259064-100MG BLDPBD259064-1G BLDPBD259064-250MG
2-Iodo-6-nitrophenol 97%
2-Iodo-6-nitrophenol

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₆H₄INO₃
Molecular Weight: 265,005 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00859244
CAS-Nummer: 13073-26-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR