Methyl-5-brom-2-(methylthio)benzoat 98%

BLDPBD257938-1GEA 77 EUR
BLDPBD257938-1G BLDPBD257938-250MG BLDPBD257938-5G
Methyl-5-brom-2-(methylthio)benzoat 98%
Methyl-5-brom-2-(methylthio)benzoat

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₉BrO₂S
Molecular Weight: 261,1356 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09258758
CAS-Nummer: 929000-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR