Methyl-4-chlor-7-methoxy-6-chinolincarboxylat 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD257784-10GEA 102 EUR
BLDPBD257784-10G BLDPBD257784-1G BLDPBD257784-250MG BLDPBD257784-5G BLDPBD257784-100G BLDPBD257784-25G
Methyl-4-chlor-7-methoxy-6-chinolincarboxylat 97%
Methyl-4-chlor-7-methoxy-6-chinolincarboxylat

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₂H₁₀ClNO₃
Molecular Weight: 251,6700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17016133
CAS-Nummer: 205448-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR