9,10-Di(naphthalen-2-yl)-2-phenylanthracene 98%

BLDPBD257783-100MGEA 66 EUR
BLDPBD257783-100MG
9,10-Di(naphthalen-2-yl)-2-phenylanthracene 98%
9,10-Di(naphthalen-2-yl)-2-phenylanthracene

Storage condition: Sealed in dry, Room Temperature

Formel: C₄₀H₂₆
Molecular Weight: 506,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16038223
CAS-Nummer: 865435-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR