4-Chloro-2-methoxypyridine-3-carbaldehyde 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD257181-1GEA 132 EUR
BLDPBD257181-1G
4-Chloro-2-methoxypyridine-3-carbaldehyde 95%
4-Chloro-2-methoxypyridine-3-carbaldehyde

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₇H₆ClNO₂
Molecular Weight: 171,581 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD13188720
CAS-Nummer: 1008451-58-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR