2,5-Bis(2-ethylhexyl)-3,6-di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione 95%

BLDPBD254710-1GEA 158 EUR
BLDPBD254710-1G BLDPBD254710-100MG BLDPBD254710-5G BLDPBD254710-250MG
2,5-Bis(2-ethylhexyl)-3,6-di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione 95%
2,5-Bis(2-ethylhexyl)-3,6-di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₃₀H₄₀N₂O₂S₂
Molecular Weight: 524,78 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22200076
CAS-Nummer: 1185885-86-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR