Methyl-4-amino-3-chlor-5-nitrobenzoat 98%

BLDPBD254190-5GEA 102 EUR
BLDPBD254190-5G BLDPBD254190-1G BLDPBD254190-250MG
Methyl-4-amino-3-chlor-5-nitrobenzoat 98%
Methyl-4-amino-3-chlor-5-nitrobenzoat

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₇ClN₂O₄
Molecular Weight: 230,6052 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 863886-04-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR