4-Methoxy-N-methylanilin Hydrochlorid 98%

BLDPBD251684-5GEA 58 EUR
BLDPBD251684-5G BLDPBD251684-10G BLDPBD251684-1G BLDPBD251684-25G BLDPBD251684-100G
4-Methoxy-N-methylanilin Hydrochlorid 98%
4-Methoxy-N-methylanilin Hydrochlorid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₁₂ClNO
Molecular Weight: 173,6400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13196471
CAS-Nummer: 10541-33-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR